missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-3-(4-pyridyl)-D-alanine, 95%
CAS: 205528-30-9 | C23H20N2O4 | 388.42 g/mol
$126.22 - $386.60
Chemical Identifiers
| CAS | 205528-30-9 |
|---|---|
| Molecular Formula | C23H20N2O4 |
| Molecular Weight (g/mol) | 388.42 |
| MDL Number | MFCD00672567 |
| InChI Key | SCSSXJVRZMQUKA-UHFFFAOYNA-N |
| Synonym | fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid |
| PubChem CID | 978323 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
| SMILES | OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52193MD
|
Thermo Scientific Chemicals
H52193MD |
250 mg |
Each for $126.22
|
|
|||||
|
AAH5219303
|
Thermo Scientific Chemicals
H5219303 |
1 g |
Each for $386.60
|
|
|||||
Description
N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 205528-30-9 | |
| 388.42 | |
| SCSSXJVRZMQUKA-UHFFFAOYNA-N | |
| 978323 | |
| OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C23H20N2O4 | |
| MFCD00672567 | |
| fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
Specifications
| 205528-30-9 | |
| MFCD00672567 | |
| fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 388.42 | |
| 388.42 | |
| N-Fmoc-3-(4-pyridyl)-D-alanine |
| C23H20N2O4 | |
| 250 mg | |
| SCSSXJVRZMQUKA-UHFFFAOYNA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid | |
| 978323 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only