Learn More
N-Fmoc-3-(4-pyridyl)-D-alanine, 95%
CAS: 205528-30-9 | C23H20N2O4 | 388.42 g/mol
Supplier: Thermo Scientific Chemicals H5219303
Description
N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| N-Fmoc-3-(4-pyridyl)-D-alanine | |
| C23H20N2O4 | |
| 1 g | |
| SCSSXJVRZMQUKA-UHFFFAOYNA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid | |
| 978323 | |
| 95% |
| 205528-30-9 | |
| MFCD00672567 | |
| fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 388.42 | |
| 388.42 |
Chemical Identifiers
| 205528-30-9 | |
| 388.42 | |
| SCSSXJVRZMQUKA-UHFFFAOYNA-N | |
| 978323 | |
| OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C23H20N2O4 | |
| MFCD00672567 | |
| fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only