accessibility menu, dialog, popup

UserName

CAS RN 205528-30-9

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-4-yl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid
Molecular Weight (g/mol): 388.42
Molecular Formula: C23H20N2O4
InChi Key: SCSSXJVRZMQUKA-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results