missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(3-pyridyl)-L-alanine, 95%

Catalog No. p-7034373
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52056MD 250 mg
AAH5205603 1 g
AAH5205606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52056MD Supplier Thermo Scientific Chemicals Supplier No. H52056MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 175453-07-3 | C23H20N2O4 | 388.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175453-07-3
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00144887
InChI Key JQLPMTXRCLXOJO-NRFANRHFSA-N
Synonym fmoc-3-3-pyridyl-l-alanine, fmoc-l-3-pyridylalanine, fmoc-3-pal-oh, fmoc-beta-3-pyridyl-ala-oh, fmoc-l-3-3-pyridyl-alanine, fmoc-3-pyridyl-ala-oh, fmoc-3'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid, n-fmoc-3-3-pyridyl-l-alanine, fmoc-d-3-3-pyridyl-alanine
PubChem CID 6957974
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O

Specifications

Quantity 250 mg
Beilstein 9154190
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(3-pyridyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only