missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(3-pyridyl)-L-alanine, 95%

CAS: 175453-07-3 | C23H20N2O4 | 388.423 g/mol

Supplier:  Thermo Scientific Chemicals H5205603

 View more versions of this product

Catalog No. AAH5205603

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-3-(3-pyridyl)-L-alanine
C23H20N2O4
1 g
fmoc-3-3-pyridyl-l-alanine, fmoc-l-3-pyridylalanine, fmoc-3-pal-oh, fmoc-beta-3-pyridyl-ala-oh, fmoc-l-3-3-pyridyl-alanine, fmoc-3-pyridyl-ala-oh, fmoc-3'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid, n-fmoc-3-3-pyridyl-l-alanine, fmoc-d-3-3-pyridyl-alanine
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O
388.423
388.42
175453-07-3
MFCD00144887
9154190
JQLPMTXRCLXOJO-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid
6957974
95%

Chemical Identifiers

175453-07-3
388.423
JQLPMTXRCLXOJO-NRFANRHFSA-N
6957974
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O
C23H20N2O4
MFCD00144887
fmoc-3-3-pyridyl-l-alanine, fmoc-l-3-pyridylalanine, fmoc-3-pal-oh, fmoc-beta-3-pyridyl-ala-oh, fmoc-l-3-3-pyridyl-alanine, fmoc-3-pyridyl-ala-oh, fmoc-3'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid, n-fmoc-3-3-pyridyl-l-alanine, fmoc-d-3-3-pyridyl-alanine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only