CAS RN 175453-07-3
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-3-yl)propanoate
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(pyridin-3-yl)propanoate
Molecular Weight (g/mol):
387.42
Molecular Formula:
C23H19N2O4
InChi Key:
JQLPMTXRCLXOJO-NRFANRHFSA-M
SMILES:
[O-]C(=O)[C@H](CC1=CC=CN=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
N-Fmoc-3-(3-pyridyl)-L-alanine, 95%
N-Fmoc-3-(3-pyridyl)-L-alanine, 95%
CAS: 175453-07-3 Molecular Formula: C23H20N2O4 Molecular Weight (g/mol): 388.423 MDL Number: MFCD00144887 InChI Key: JQLPMTXRCLXOJO-NRFANRHFSA-N Synonym: fmoc-3-3-pyridyl-l-alanine,fmoc-l-3-pyridylalanine,fmoc-3-pal-oh,fmoc-beta-3-pyridyl-ala-oh,fmoc-l-3-3-pyridyl-alanine,fmoc-3-pyridyl-ala-oh,fmoc-3'-pyridyl-l-ala,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid,n-fmoc-3-3-pyridyl-l-alanine,fmoc-d-3-3-pyridyl-alanine PubChem CID: 6957974 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid SMILES: C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O