missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, 98+%

Numéro de catalogue. p-7058292
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0437206 5 g
AAL0437214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0437206 Fournisseur Thermo Scientific Chemicals Code fournisseur L0437206
Il en reste null

CAS: 624-78-2 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
PubChem CID 12219
IUPAC Name N-methylethanamine
SMILES CCNC

Spécifications

Density 0.688
Boiling Point 33°C to 34°C
Flash Point <−34°C (−29°F)
Odor Amine-like
Refractive Index 1.374
Quantity 5 g
UN Number UN2733
Beilstein 1730786
Sensitivity Air sensitive; Hygroscopic
Formula Weight 59.11
Percent Purity ≥98%
Chemical Name or Material N-Ethylmethylamine
Afficher plus de résultats Afficher moins de résultats
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-862-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only