missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylethylenediamine 99.0+%, TCI America™

Catalog No. p-7128918
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E029425ML 25 mL
E0294100ML 100 mL
E0294500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E029425ML Supplier TCI America Supplier No. E029425ML

Chemical Identifiers

CAS 110-72-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008166
InChI Key SCZVXVGZMZRGRU-UHFFFAOYSA-N
Synonym n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
PubChem CID 66071
IUPAC Name N'-ethylethane-1,2-diamine
SMILES CCNCCN

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 2734
Formula Weight 88.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethylethylenediamine
EINECSNumber (2)-3275
TSCA Yes