missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylbenzylamine 97.0+%, TCI America™

Supplier:  TCI America E0383100ML

 View more versions of this product

Catalog No. E0383100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Ethylbenzylamine
Yellow
C9H13N
100 mL
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
CCNCC1=CC=CC=C1
135.21
135.21
Liquid
14321-27-8
199°C
MFCD00009031
2735
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
N-benzylethanamine
84352
≥97.0% (GC,T)

Chemical Identifiers

14321-27-8
135.21
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
84352
CCNCC1=CC=CC=C1
C9H13N
MFCD00009031
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
N-benzylethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0231

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions