accessibility menu, dialog, popup

UserName

H3CHN

CAS RN 14321-27-8

IUPAC Name: benzyl(ethyl)amine
Synonyms: benzenemethanamine, N-ethyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: HVAAHUDGWQAAOJ-UHFFFAOYSA-N
SMILES: CCNCC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)