missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylbenzylamine 97.0+%, TCI America™

Catalog No. p-7128973
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E038325ML 25 mL
E0383100ML 100 mL
E0383500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E038325ML Supplier TCI America Supplier No. E038325ML
Only null left

Chemical Identifiers

CAS 14321-27-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00009031
InChI Key HVAAHUDGWQAAOJ-UHFFFAOYSA-N
Synonym n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
PubChem CID 84352
IUPAC Name N-benzylethanamine
SMILES CCNCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 mL
UN Number 2735
Formula Weight 135.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethylbenzylamine
EINECSNumber (9)-0231
TSCA Yes