missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylaniline 99.0+%, TCI America™

Numéro de catalogue. p-7128813
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E005925ML 25 mL
E0059500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E005925ML Fournisseur TCI America Code fournisseur E005925ML
Il en reste null

Identifiants chimiques

CAS 103-69-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00009025
InChI Key OJGMBLNIHDZDGS-UHFFFAOYSA-N
Synonym ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
PubChem CID 7670
ChEBI CHEBI:34870
IUPAC Name N-ethylaniline
SMILES CCNC1=CC=CC=C1

Spécifications

Melting Point -63°C
Color Yellow
Boiling Point 204°C
Quantity 25 mL
UN Number 2272
Formula Weight 121.18
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethylaniline
EINECSNumber (3)-0118&(3)-0206
RTECSNumber BX9780000
TSCA Yes