missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-toluidine 98.0+%, TCI America™

Catalog No. E017925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017925ML Supplier TCI America Supplier No. E017925ML

Specifications

CAS 622-57-1
Color Yellow
Boiling Point 222°C
Molecular Formula C9H13N
Quantity 25 mL
MDL Number MFCD00035793
UN Number 2754
Synonym n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
InChI Key AASABFUMCBTXRL-UHFFFAOYSA-N
SMILES CCNC1=CC=C(C=C1)C
IUPAC Name N-ethyl-4-methylaniline
Molecular Weight (g/mol) 135.21
PubChem CID 61164
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethyl-p-toluidine
Show More Show Less
EINECSNumber (3)-0187&(3)-0193
TSCA Yes