missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(4-picolyl)atropamide 90.0+%, TCI America™

Catalog No. E0502100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0502100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0502100MG Supplier TCI America Supplier No. E0502100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57322-50-6
Molecular Formula C17H18N2O
Molecular Weight (g/mol) 266.34
MDL Number MFCD01321167
InChI Key LOVSQYAVWGMIRV-UHFFFAOYSA-N
Synonym N-Ethyl-N-(4-picolyl)-2-phenylacrylamide, N-Ethyl-N-(4-pyridylmethyl)-2-phenylacrylamide
PubChem CID 44630282
IUPAC Name N-ethyl-2-phenyl-N-[(pyridin-4-yl)methyl]prop-2-enamide
SMILES CCN(CC1=CC=NC=C1)C(=O)C(=C)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 266.34
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-(4-picolyl)atropamide