missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-2-hydroxyethyl-m-toluidine 96.0+%, TCI America™

Catalog No. E0220500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0220500G 500 g
E022025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0220500G Supplier TCI America Supplier No. E0220500G
Only null left

Chemical Identifiers

CAS 91-88-3
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.263
MDL Number MFCD00002849
InChI Key KRNUKKZDGDAWBF-UHFFFAOYSA-N
Synonym 2-n-ethyl-m-toluidino ethanol, 2-ethyl m-tolyl amino ethanol, emery 5714, n-ethyl-n-2-hydroxyethyl-m-toluidine, ethanol, 2-ethyl 3-methylphenyl amino, ethanol, 2-n-ethyl-m-toluidino, n-ethyl-n-hydroxyethyl-meta-toluidine, n-hydroxyethyl-n-ethyl-m-toluidine, unii-1alr5x8r8w
PubChem CID 7067
IUPAC Name 2-(N-ethyl-3-methylanilino)ethanol
SMILES CCN(CCO)C1=CC=CC(=C1)C

Specifications

Boiling Point 147°C
Quantity 500 g
Formula Weight 179.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-2-hydroxyethyl-m-toluidine
EINECSNumber (3)-0203
RTECSNumber KL1320000
TSCA Yes