missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Dodecylaniline 93.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 261.45 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | N-Dodecylaniline |
Sécurité et manipulation
| TSCA | No |