missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Dodecyl-β-D-maltoside, ULTROL™, ≥99%, Calbiochem™,

Catalog No. 3243555GM
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3243555GM 5 g
32435525GM 25 g
324355100GM 100 g
324355250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3243555GM Supplier MilliporeSigma™ Supplier No. 3243555GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color White
Molecular Formula C24H46O11
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 510.621
PubChem CID 114880
ChEBI CHEBI:43769
Quantity 5 g
Percent Purity ≥98% (HPLC)
Format Glass Bottle
Show More Show Less