missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, EMD-Millipore

Catalog No. M1096030005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096030005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096030005 Supplier Technidata Supplier No. 1.09603.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrocarbons C9 – C19

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 0.7 to 5.4 %(V)
Ignition Point 208°C
Vapor Pressure 1 hPa (16°C)
Viscosity <7mm2/sec., kinematic (40°C)
Density Density: 0.73g/cm3 (20°)
Assay Percent Range ≥99.5%
Linear Formula CH3(CH2)8CH3
Quantity 5 mL
Formula Weight 142.28
Chemical Name or Material n-Decane
Show More Show Less
Recommended Storage Store without restrictions