missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, 99%, Spectrum™ Chemical

Catalog No. 18611920
Change view
Click to view available options
Quantity:
100 mL
500 mL
13 kg
150 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611920 500 mL Amber Glass Bottle
18611917 100 mL Amber Glass Bottle
18611918 13 kg Amber Glass Bottle
18611919 150 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611920 Supplier Spectrum Chemical Mfg Cor Supplier No. D2098500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-18-5

Specifications

CAS Max % 1
Boiling Point 174.1°C/345.4°F
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 0.99
Grade Ungraded
Physical Form Liquid
Chemical Name or Material n-Decane

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.