missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Chlorosuccinimide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 148°C |
| Color | White |
| Quantity | 100 g |
| UN Number | 3261 |
| Formula Weight | 133.53 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Chlorosuccinimide |
Safety and Handling
| EINECSNumber | (5)-0121 |
| RTECSNumber | UY1013500 |
| TSCA | Yes |