missing translation for 'onlineSavingsMsg'
Learn More

N-Chloromethylphthalimide 98.0+%, TCI America™

Catalog No. A55045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55045G Supplier TCI America Supplier No. A55045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17564-64-6
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00005898
InChI Key JKGLRGGCGUQNEX-UHFFFAOYSA-N
Synonym n-chloromethyl phthalimide, 2-chloromethyl isoindoline-1,3-dione, n-chloromethylphthalimide, chloromethylphthalimide, n-chloromethyltrimellitimide, 1h-isoindole-1,3 2h-dione, 2-chloromethyl, phthalimide, n-chloromethyl, 2-chloromethyl isoindole-1,3-dione, 2-chloromethyl-1h-isoindole-1,3 2h-dione
PubChem CID 87154
IUPAC Name 2-(chloromethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCl

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 195.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloromethylphthalimide [for HPLC Labeling]