missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxyhydroxylamine 97.0+%, TCI America™

Catalog No. C15575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15575G 5 g
C155725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15575G Supplier TCI America Supplier No. C15575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3426-71-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00010642
InChI Key PQBSPTAPCMSZAA-UHFFFAOYSA-N
Synonym benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
PubChem CID 18907
IUPAC Name benzyl N-hydroxycarbamate
SMILES ONC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxyhydroxylamine
RTECSNumber FB1600000
TSCA No
Recommended Storage Refrigerator