missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-tryptophan 98.0+%, TCI America™

Catalog No. C06385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06385G 5 g
C063825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06385G Supplier TCI America Supplier No. C06385G

Chemical Identifiers

CAS 7432-21-5
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00065700
InChI Key AHYFYYVVAXRMKB-KRWDZBQOSA-N
Synonym z-trp-oh, n-cbz-l-tryptophan, z-l-trp-oh, benzyloxycarbonyltryptophan, l-tryptophan, n-phenylmethoxy carbonyl, n-carbobenzoxyltryptophan, n-carbobenzoxy-l-tryptophan, cbz-l-trp-oh, n-benzyloxycarbonyl-l-tryptophan, nalpha-carbobenzyloxy-l-tryptophan
PubChem CID 101176
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-tryptophan