missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-threonine 98.0+%, TCI America™

Catalog No. C135110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C135110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C135110G Supplier TCI America Supplier No. C135110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19728-63-3
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00065948
InChI Key IPJUIRDNBFZGQN-SCZZXKLOSA-N
Synonym z-thr-oh, n-cbz-l-threonine, z-l-threonine, z-l-thr-oh, n-benzyloxycarbonyl-l-threonine, cbz-l-threonine, n-carbobenzyloxy-l-threonine, l-threonine, n-phenylmethoxy carbonyl, benzyloxycarbonyl-l-threonine, n-benzyloxy carbonyl-l-threonine
PubChem CID 88217
IUPAC Name (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid
SMILES C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 103°C
Color White
Quantity 10 g
Formula Weight 253.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-threonine
EINECSNumber (3)-1027
TSCA No