missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-phénylalanine 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 86°C |
| Color | Blanc |
| Quantity | 5 g |
| Formula Weight | 299.33 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | N-Carbobenzoxy-L-phenylalanine |
Sécurité et manipulation
| RTECSNumber | AY4343000 |
| TSCA | No |