missing translation for 'onlineSavingsMsg'
Learn More

N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine 98.0+%, TCI America™

Numéro de catalogue. C27091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C27091G 1 g
C27095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C27091G Fournisseur TCI America Code fournisseur C27091G
Il en reste null

Identifiants chimiques

CAS 152120-62-2
Molecular Formula C12H12N4O2
Molecular Weight (g/mol) 244.254
MDL Number MFCD00274657
InChI Key BXUMISWXFKCEIA-UHFFFAOYSA-N
Synonym n-z-1h-pyrazole-1-carboxamidine, benzyl imino 1h-pyrazol-1-yl methyl carbamate, n-benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n-carbobenzoxy-1h-pyrazole-1-carboxamidine, n-z-1h-pyrazole-1-carboxamdne, benzyl n-imino 1-pyrazolyl methyl carbamate, benzyl imino-1h-pyrazol-1-yl methylcarbamate
PubChem CID 11857377
IUPAC Name benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
SMILES C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2

Spécifications

Melting Point 111°C
Color White
Quantity 1 g
Formula Weight 244.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine