missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,4-diaminobutane Hydrochloride 97.0+%, TCI America™

Catalog No. C15195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15195G 5 g
C15191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15195G Supplier TCI America Supplier No. C15195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18807-73-3
Molecular Formula C12H19ClN2O2
Molecular Weight (g/mol) 258.75
MDL Number MFCD00270149
InChI Key ZVNNCIIFBSRHFE-UHFFFAOYSA-N
Synonym N-(4-Aminobutyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(4-Aminobutyl)carbamate Hydrochloride, N-Cbz-1,4-diaminobutane Hydrochloride
PubChem CID 13120301
IUPAC Name benzyl N-(4-aminobutyl)carbamate hydrochloride
SMILES Cl.NCCCCNC(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 258.75
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,4-diaminobutane Hydrochloride