Learn More
n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol, Thermo Scientific Chemicals
Description
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.
Solubility
Miscible with ethyl ether and ethanol. Immiscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.
Specifications
Specifications
| Melting Point | -75°C |
| Density | 0.894 |
| Boiling Point | 160°C to 163°C |
| Flash Point | 49°C (120°F) |
| Odor | Fruit-like |
| Refractive Index | 1.424 |
| Quantity | 500 mL |
| UN Number | UN2227 |
| Beilstein | 773960 |
| Solubility Information | Miscible with ethyl ether and ethanol. Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H331-H226-H315-H319-H317-H335 Toxic if inhaled. Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-BUTYL METHACRYLATE, STABILIZED |
| EINECSNumber | 202-615-1 |
| RTECSNumber | OZ3675000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only