missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-trans-4-N-FMOC-amino-L-proline, 97%

Catalog No. AC426040010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426040010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426040010 Supplier Thermo Scientific Chemicals Supplier No. 426040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 176486-63-8 | C25H28N2O6 | 452.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 176486-63-8
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.5
MDL Number MFCD00673783
InChI Key UPXRTVAIJMUAQR-VFNWGFHPSA-N
Synonym 2s,4r-4-9h-fluoren-9-yl methoxy carbonyl amino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-boc-trans-4-n-fmoc-amino-l-proline, 2s,4r-n-tert-butoxycarbonyl-4-n-9-fluorenylmethoxycarbonyl aminopyrrolidine-2-carboxylic acid, 2s,4r-4-9h-fluoren-9-yl methoxy carbonylamino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino pyrrolidine-2-carboxylic acid, ambotzbaa1783, 2s,4r-fmoc-4-amino-1-boc-pyrrolidine-2-carboxyli, 2s,4r-1-boc-4-n-fmoc-4-aminopyrrolidine-2-carboxylic acid, 2s,4r-trans-n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-proline
PubChem CID 2756135
IUPAC Name (2S,4R)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 199°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -33° to -35° (20°C, 589nm) (c=1, chloroform)
Formula Weight 452.5
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-trans-4-N-FMOC-amino-L-proline
Show More Show Less
Hazard Category Warning

RUO – Research Use Only