missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-L-proline methyl ester, 97%, Thermo Scientific™

Catalog No. AAH5902703
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5902703 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5902703 Supplier Thermo Scientific Chemicals Supplier No. H5902703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis of For-Met-Leu-Phe-OMe (fMLF-OMe) analogues based on Met residue replacement by 4-amino-proline scaffold.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications

CAS 74844-91-0
Molecular Formula C11H19NO5
Quantity 1g
MDL Number MFCD00076981
Synonym boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
InChI Key MZMNEDXVUJLQAF-SFYZADRCSA-N
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
Optical Rotation −65° (c=1 in Chloroform)
IUPAC Name 1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol) 245.28
PubChem CID 2734883
Formula Weight 245.28
Percent Purity 97%
Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline methyl ester
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only