missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-valine, 95%

Catalog No. AAH6277106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6277106 5 g
AAH6277103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6277106 Supplier Thermo Scientific Chemicals Supplier No. H6277106
Only null left

CAS: 45170-31-8 | C11H21NO4 | 231.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 45170-31-8
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038760
InChI Key XPUAXAVJMJDPDH-QMMMGPOBSA-N
Synonym boc-n-me-val-oh, boc-n-methyl-l-valine, n-boc-n-methyl-l-valine, boc-meval-oh, s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-methylbutanoic acid, ambotzbaa1272, n-boc-n-methylvaline, pubchem12254
PubChem CID 7010608
IUPAC Name (2S)-3-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]butanoic acid
SMILES CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

Specifications

Beilstein 2646180
Quantity 5 g
Formula Weight 231.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-valine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only