missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-D-phenylalanine, 98%

Catalog No. AAH62638MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62638MD 250 mg
AAH6263803 1 g
AAH6263806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62638MD Supplier Thermo Scientific Chemicals Supplier No. H62638MD
Only null left

CAS: 85466-66-6 | C15H21NO4 | 279.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85466-66-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00151890
InChI Key AJGJINVEYVTDNH-UHFFFAOYNA-N
Synonym boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid
PubChem CID 7015127
SMILES CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C

Specifications

Color White
IUPAC Name (2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid
Quantity 250 mg
Formula Weight 279.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-N-methyl-D-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only