missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-valine, 98+%

Catalog No. AAA1600714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1600714 25 g
AAA1600706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1600714 Supplier Thermo Scientific Chemicals Supplier No. A1600714

CAS: 13734-41-3 | C10H19NO4 | 217.27 g/mol

N-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13734-41-3
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00065605
InChI Key SZXBQTSZISFIAO-ZETCQYMHSA-N
Synonym boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
PubChem CID 83693
IUPAC Name (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 77°C to 80°C
Quantity 25 g
Beilstein 1711290
Solubility Information Insoluble in water.
Optical Rotation −7.5° (c=1 in Acetic Acid)
Formula Weight 217.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-valine
EINECSNumber 237-307-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only