missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-BOC-L-Tryptophane, 97%

Catalog No. AC275691000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275691000 100 g Glass Bottle
AC275690050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275691000 Supplier Thermo Scientific Chemicals Supplier No. 275691000

CAS: 13139-14-5 | C16H20N2O4 | 304.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13139-14-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD000655
InChI Key NFVNYBJCJGKVQK-ZDUSSCGKSA-N
Synonym boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
PubChem CID 83169
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Beilstein 22, 6789
Specific Rotation -19° to -23° (20°C, 589nm) (c=1, CH3COOH)
Water 1% Max. (K.F.)
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Specific Rotation Condition −20° (20°C c=1,methanol)
Quantity 100 g
Formula Weight 304.34
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Nα-BOC-L-Tryptophane
Show More Show Less

RUO – Research Use Only