missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-proline methyl ester, 96%

Catalog No. p-7058156
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6236314 25 g
AAH6236322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6236314 Supplier Thermo Scientific Chemicals Supplier No. H6236314

CAS: 59936-29-7 | C11H19NO4 | 229.28 g/mol

N-Boc-L-proline methyl ester is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-proline methyl ester is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 59936-29-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01075092
InChI Key WVDGSSCWFMSRHN-QMMMGPOBSA-N
Synonym boc-l-pro-ome, boc-pro-ome, boc-l-proline-methyl ester, s-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, boc-l-proline methyl ester, 1-tert-butyl 2-methyl 2s-pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl s-pyrrolidine-1,2-dicarboxylate, pubchem18390, boc-l-prolinemethylester, boc-l-prolinem ethylester
PubChem CID 1277666
SMILES COC(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-pyrrolidine-1,2-dicarboxylate
Formula Weight 229.27
Percent Purity 96%
Chemical Name or Material N-Boc-L-proline methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only