missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanine, 99%

Catalog No. AAA1601714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1601714 25 g
AAA1601706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1601714 Supplier Thermo Scientific Chemicals Supplier No. A1601714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

Solubility
Soluble in methanol, dichloromethane, dimethylformamide and N-methyl-2-pyrrolidone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 84°C to 88°C
Quantity 25 g
Beilstein 2219729
Solubility Information Soluble in methanol,dichloromethane,dimethylformamide and N-methyl-2-pyrrolidone.
Optical Rotation +25° (c=1 in Ethanol)
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
Formula Weight 265.31
Percent Purity 99%
Chemical Name or Material N-Boc-L-phenylalanine
EINECSNumber 237-305-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only