missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals

Numéro de catalogue. AAA1064706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1064706 5 g
AAA1064714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1064706 Fournisseur Thermo Scientific Chemicals Code fournisseur A1064706
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 13139-15-6
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00066067
InChI Key MDXGYYOJGPFFJL-SVGMAFHSNA-N
Synonym boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine
PubChem CID 83170
ChEBI CHEBI:28702
IUPAC Name (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Spécifications

Melting Point 84°C to 87°C
Quantity 5 g
Beilstein 1711814
Optical Rotation −25° (c=2 in acetic acid)
Formula Weight 231.29 (Anhydrous)
Percent Purity 99%
Chemical Name or Material N-Boc-L-leucine hydrate
EINECSNumber 236-073-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only