missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alaninamide, 96%

Catalog No. AAH62604MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62604MD 250 mg
AAH6260403 1 g
AAH6260406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62604MD Supplier Thermo Scientific Chemicals Supplier No. H62604MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85642-13-3 | C8H16N2O3 | 188.23 g/mol

N-Boc-L-alaninamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-alaninamide is used as pharmaceutical intermediate.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 85642-13-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD03701447
InChI Key GZKKSKKIABUUCX-UHFFFAOYNA-N
Synonym boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310
PubChem CID 7015295
SMILES CC(NC(=O)OC(C)(C)C)C(N)=O

Specifications

Quantity 250 mg
Solubility Information Soluble in water or 1% acetic acid.
IUPAC Name tert-butyl N-(1-carbamoylethyl)carbamate
Formula Weight 188.22
Percent Purity 96%
Physical Form Solid
Chemical Name or Material N-Boc-L-alaninamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only