missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine ethyl ester, 95%

Catalog No. p-7054417
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6261106 5 g
AAH6261114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6261106 Supplier Thermo Scientific Chemicals Supplier No. H6261106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14719-37-0 | C9H17NO4 | 203.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14719-37-0
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD05663720
InChI Key CNIBHMMDDXGDNR-UHFFFAOYSA-N
Synonym n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
PubChem CID 357729
IUPAC Name ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CCOC(=O)CNC(=O)OC(C)(C)C

Specifications

Color Colorless
Linear Formula (CH3)3COCONHCH2CO2CH2CH3
Quantity 5 g
Formula Weight 203.24
Percent Purity 95%
Physical Form Oily Liquid
Chemical Name or Material N-Boc-glycine ethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only