missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-threonine, 95%

Catalog No. AAH6364714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6364714 25 g
AAH6364706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6364714 Supplier Thermo Scientific Chemicals Supplier No. H6364714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55674-67-4 | C9H17NO5 | 219.24 g/mol

It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 55674-67-4
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
MDL Number MFCD00037807
InChI Key LLHOYOCAAURYRL-NTSWFWBYSA-N
Synonym boc-d-thr-oh, boc-d-threonine, n-tert-butoxycarbonyl-d-threonine, n-boc-d-threonine, 2r,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid, n-alpha-tert-butoxycarbonyl-d-threonine, d-threonine, n-1,1-dimethylethoxy carbonyl, pubchem14945, boc-d-thr
PubChem CID 6992541
IUPAC Name (2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Beilstein 2051734
Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 219.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-D-threonine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only