missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-phenylalanine, 98%

Catalog No. AAL0872206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0872206 5 g
AAL0872203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0872206 Supplier Thermo Scientific Chemicals Supplier No. L0872206
Only null left

CAS: 18942-49-9 | C14H19NO4 | 265.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18942-49-9
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00063149
InChI Key ZYJPUMXJBDHSIF-LLVKDONJSA-N
Synonym boc-d-phenylalanine, boc-d-phe-oh, n-boc-d-phenylalanine, n-tert-butoxycarbonyl-d-phenylalanine, boc-d-phe, d-phenylalanine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-phenylalanine, 2r-2-tert-butoxy carbonylamino-3-phenylpropanoic acid, 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoic acid
PubChem CID 637610
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
Beilstein 3593396
Optical Rotation −24.5° (c=1 in Ethanol)
Formula Weight 265.31
Percent Purity 98%
Chemical Name or Material N-Boc-D-phenylalanine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only