missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid, 98%

Numéro de catalogue. AAH6276206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH6276206 5 g
AAH6276214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH6276206 Fournisseur Thermo Scientific Chemicals Code fournisseur H6276206
Il en reste null

CAS: 34404-28-9 | C10H17NO6 | 247.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 34404-28-9
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.247
MDL Number MFCD00190790
InChI Key AQTUACKQXJNHFQ-ZCFIWIBFSA-N
Synonym boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid
PubChem CID 7019046
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O

Spécifications

Quantity 5 g
Formula Weight 247.25
Percent Purity 98%
Chemical Name or Material N-Boc-D-glutamic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only