missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alanine, 98+%

Catalog No. AAB2270606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2270606 5 g
AAB2270614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2270606 Supplier Thermo Scientific Chemicals Supplier No. B2270606

CAS: 7764-95-6 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7764-95-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00063123
InChI Key QVHJQCGUWFKTSE-RXMQYKEDSA-N
Synonym boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 637606
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES C[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 81°C to 84°C
Quantity 5 g
Beilstein 2048396
Optical Rotation +26° (c=2 in Acetic Acid)
Formula Weight 189.21
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only