missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-L-proline methyl ester, 97%, Thermo Scientific™

Catalog No. AAH2701806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2701806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2701806 Supplier Thermo Scientific Chemicals Supplier No. H2701806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a local anaesthetic. Used as therapeutic agents. Employed as important intermediates in various areas such as peptide synthesis, polymer chemistry..

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 102195-79-9
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD00237541
InChI Key MZMNEDXVUJLQAF-YUMQZZPRSA-N
Synonym n-boc-cis-4-hydroxy-l-proline methyl ester, boc-cis-hyp-ome, 2s,4s-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, cis-boc-hyp-ome, boc-cis-4-hydroxy-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-l-prolinate, 2s,4s-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, methyl 4s-n-boc-4-hydroxy-l-prolinate, o1-tert-butyl o2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate
PubChem CID 10752873
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4S)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

Specifications

Melting Point 81°C to 84°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 245.28
Percent Purity 97%
Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline methyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold