missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-methyl-L-phenylalanine, 95%

Catalog No. p-7034312
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5198303 1 g
AAH5198306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5198303 Supplier Thermo Scientific Chemicals Supplier No. H5198303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80102-26-7 | C15H21NO4 | 279.34 g/mol

It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 80102-26-7
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD01317027
InChI Key JYRWNPUFECDJCX-LBPRGKRZSA-N
Synonym n-boc-4-methyl-l-phenylalanine, boc-phe 4-me-oh, boc-l-4-methylphe, boc-4-methyl-l-phenylalanine, boc-l-4-methylphenylalanine, boc-4-methy-l-phenylalanine, boc-p-me-phe-oh, boc-l-4-me-phe-oh, 2s-2-tert-butoxycarbonyl amino-3-4-methylphenyl propanoic acid, l-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-methyl
PubChem CID 2761472
SMILES CC1=CC=C(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Melting Point 84°C to 88°C
Quantity 1 g
Beilstein 5381557
Solubility Information Slightly soluble in water.
Optical Rotation +16° (c=1 in Methanol)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid
Formula Weight 279.34
Percent Purity 95%
Chemical Name or Material N-Boc-4-methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only