missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-cyano-L-phenylalanine, 95%

Catalog No. AAH6202303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6202303 1 g
AAH6202306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6202303 Supplier Thermo Scientific Chemicals Supplier No. H6202303

CAS: 131724-45-3 | C15H18N2O4 | 290.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131724-45-3
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD00672526
InChI Key RMBLTLXJGNILPG-LBPRGKRZSA-N
Synonym boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh
PubChem CID 7020841
IUPAC Name (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O

Specifications

Melting Point 156°C to 160°C
Color White
UN Number UN3439
Beilstein 7715793
Quantity 1 g
Formula Weight 290.31
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-4-cyano-L-phenylalanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only