missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-dimethylamino-DL-alanine, 97%

Catalog No. AAH5241203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5241203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5241203 Supplier Thermo Scientific Chemicals Supplier No. H5241203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 851653-36-6 | C10H20N2O4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 851653-36-6
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD04972270
InChI Key VCDQZVYJKDSORW-UHFFFAOYSA-N
Synonym 2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, nalpha-boc-+/--2-amino-3-dimethylamino propionic acid, n-tert-butoxycarbonyl-3-dimethylamino alanine, n?-boc-2-amino-3-dimethylamino propionic acid, alanine,3-dimethylamino-n-1,1-dimethylethoxy c, 2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, 2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-alpha-boc-+/--2-amino-3-dimethylamino propionic acid
PubChem CID 13850981
IUPAC Name 3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O

Specifications

Quantity 1 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material N-Boc-3-dimethylamino-DL-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only