missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-bromo-L-phenylalanine, 95%

Catalog No. AAH5202406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5202406 5 g
AAH5202403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5202406 Supplier Thermo Scientific Chemicals Supplier No. H5202406
Only null left

CAS: 82278-73-7 | C14H18BrNO4 | 344.205 g/mol

N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 82278-73-7
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01317710
InChI Key FBUDYESOPLBQIR-NSHDSACASA-N
Synonym s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid
PubChem CID 2734476
IUPAC Name (2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 344.2
Percent Purity 95%
Chemical Name or Material N-Boc-3-bromo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only