missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,4-dichloro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog No. AAH5205303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5205303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5205303 Supplier Thermo Scientific Chemicals Supplier No. H5205303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80741-39-5
Molecular Formula C14H16Cl2NO4
Molecular Weight (g/mol) 333.19
MDL Number MFCD00273436
InChI Key UGZIQCCPEDCGGN-NSHDSACASA-M
Synonym boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine
PubChem CID 2734501
IUPAC Name (2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O

Specifications

Quantity 1 g
Beilstein 6426101
Formula Weight 334.2
Percent Purity 95%
Chemical Name or Material N-Boc-3,4-dichloro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only