missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-methyl-L-serine, 97%

Catalog No. AAH52570MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52570MD 250 mg
AAH5257003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52570MD Supplier Thermo Scientific Chemicals Supplier No. H52570MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84311-19-3 | C9H17NO5 | 219.24 g/mol

N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with reducing agents, water and strong oxidizing agents.

Chemical Identifiers

CAS 84311-19-3
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
MDL Number MFCD02682593
InChI Key FWRXDSRYWWYTPD-VIFPVBQESA-N
Synonym n-boc-alpha-methyl-l-serine, s-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, boc-alpha-methyl-l-ser, 2s-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, s-2-boc-amino-3-hydroxy-2-methylpropionic acid, n-1,1-dimethylethyl oxy carbonyl-2-methylserine, serine, n-1,1-dimethylethoxy carbonyl-2-methyl, s-2-tert-butoxycarbonylamino-3-hydroxy-2-methylpropanoic acid, n-boc-a-methylserine, boc-a-methyl-l-ser
PubChem CID 12991620
IUPAC Name (2S)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@](C)(CO)C(O)=O

Specifications

Quantity 250 mg
Solubility Information Soluble in dimethylformamide.
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material N-Boc-2-methyl-L-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only